missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethylcyclohexanecarboxylic acid, cis + trans, 97%

Manufacturer:  Alfa Aesar™ H5500206

 View more versions of this product

Catalog No. AAH5500206



Specifications

Specifications

4-Ethylcyclohexanecarboxylic acid
Cis + Trans
C9H16O2
1s,4r-4-ethylcyclohexane-1-carboxylic acid
CCC1CCC(CC1)C(=O)O
156.225
156.23
5g
91328-77-7
MFCD01568857
UNROFSAOTBVBBT-UHFFFAOYSA-N
4-ethylcyclohexane-1-carboxylic acid
1490288
97%

Chemical Identifiers

91328-77-7
156.225
UNROFSAOTBVBBT-UHFFFAOYSA-N
1490288
CCC1CCC(CC1)C(=O)O
C9H16O2
MFCD01568857
1s,4r-4-ethylcyclohexane-1-carboxylic acid
4-ethylcyclohexane-1-carboxylic acid
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents