Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethylbiphenyl, 99%

$56.50 - $212.42

Chemical Identifiers

Chemical Name or Material 4-Ethylbiphenyl
CAS 5707-44-8
Molecular Formula C14H14
Formula Weight 182.27
MDL Number MFCD00010236
Beilstein 1904973
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2427006

 
alfa aesar™
B2427006
5g Each for $56.50

AAB2427014

 
alfa aesar™
B2427014
25g Each for $212.42
Description & Specifications

Specifications

Formula Weight 182.27
Chemical Name or Material 4-Ethylbiphenyl
Assay 99%
Melting Point 34° to 36°C
Boiling Point 140°C (15mmHg)
Molecular Formula C14H14
Quantity 5g
MDL Number MFCD00010236
CAS 5707-44-8
Beilstein 1904973