Please login to your online account to display your discounted pricing

4-Ethylbenzylamine 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Ethylbenzylamine
CAS 7441-43-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H13N
Linear Formula CH3CH2C6H4CH2NH2
Formula Weight 135.21
MDL Number MFCD01529871
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC391140010

  SDS
thermo fisher scientific
AC391140010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC391140050

  SDS
thermo fisher scientific
AC391140050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 135.21
Quantity 1g
Color Clear colorless
Density 0.8830g/mL
Beilstein 12, IV, 2641
CAS Max % 100.0
Refractive Index 1.5280 to 1.5300 (20°C, 589nm)
Packaging Glass bottle
Assay Percent Range 98%
CAS 7441-43-2
Chemical Name or Material 4-Ethylbenzylamine
Molecular Formula C9H13N
MDL Number MFCD01529871
Infrared Spectrum Authentic
Linear Formula CH3CH2C6H4CH2NH2
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.