missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Ethylbenzyl Alcohol, TCI America™

Manufacturer:  TCI America E070425G

 View more versions of this product

Catalog No. E070425G



Specifications

Specifications

4-Ethylbenzyl Alcohol
Yellow
768-59-2
MFCD00004666
YSLBFFIVJGJBSA-UHFFFAOYSA-N
(4-ethylphenyl)methanol
13034
Clear Liquid at 20°C
117°C
25g
C9H12O
4-ethylphenyl methanol, 4-ethylbenzyl alcohol, 4-ethylbenzylalcohol, benzenemethanol, 4-ethyl, benzyl alcohol, p-ethyl, p-ethylbenzyl alcohol, 4-ethyl-benzyl alcohol, benzenemethanol,4-ethyl, acmc-209p6g, 4-ethylphenyl methanol #
CCC1=CC=C(C=C1)CO
136.194
136.19
≥98.0% (GC)

Chemical Identifiers

768-59-2
136.194
YSLBFFIVJGJBSA-UHFFFAOYSA-N
13034
CCC1=CC=C(C=C1)CO
C9H12O
MFCD00004666
4-ethylphenyl methanol, 4-ethylbenzyl alcohol, 4-ethylbenzylalcohol, benzenemethanol, 4-ethyl, benzyl alcohol, p-ethyl, p-ethylbenzyl alcohol, 4-ethyl-benzyl alcohol, benzenemethanol,4-ethyl, acmc-209p6g, 4-ethylphenyl methanol #
(4-ethylphenyl)methanol
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1049

TSCA : No

Documents