Please login to your online account to display your discounted pricing

4-Ethylbenzoyl chloride, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  305730050

 View more versions of this product

Catalog No. AC305730050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 168.62
Quantity 5g
Color Clear light yellow to yellow
Density 1.14g/mL
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Linear Formula CH5C6H4COCl
CAS 16331-45-6
Chemical Name or Material 4-Ethylbenzoyl chloride
Molecular Formula C9H9ClO
Health Hazard 1 GHS Signal Word: Danger
Packaging Glass bottle
MDL Number MFCD00000697
Refractive Index 1.5465 to 1.5485 (20°C, 589nm)
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

Specifications

Chemical Name or Material 4-Ethylbenzoyl chloride
CAS 16331-45-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H9ClO
Linear Formula CH5C6H4COCl
Formula Weight 168.62
MDL Number MFCD00000697
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

SDS & Safety Documents