Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethylbenzophenone, 96%

$238.59 - $238.59

Chemical Identifiers

Chemical Name or Material 4-Ethylbenzophenone
CAS 18220-90-1
Molecular Formula C15H14O
Formula Weight 210.28
MDL Number MFCD00015177
Beilstein 1949192
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2448418

  SDS
alfa aesar™
B2448418
50g Each for $238.59
Description & Specifications

Specifications

Formula Weight 210.28
Chemical Name or Material 4-Ethylbenzophenone
Assay 96%
Boiling Point 150°C (0.5mmHg)
Molecular Formula C15H14O
Quantity 50g
Flash Point >110°C (230°F)
MDL Number MFCD00015177
CAS 18220-90-1
Beilstein 1949192

Applications
It is used in pharmaceutical industry.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.