missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethylbenzoic acid, 99%

Manufacturer:  Alfa Aesar™ A1023530

 View more versions of this product

Catalog No. AAA1023530



Specifications

Specifications

4-Ethylbenzoic acid
250g
C9H10O2
2041840
ZQVKTHRQIXSMGY-UHFFFAOYSA-N
4-ethylbenzoic acid
12086
99%
111°C to 114°C
619-64-7
MFCD00002570
benzoic acid, 4-ethyl, p-ethylbenzoic acid, 4-ethyl benzoic acid, 4-ethyl-benzoic acid, unii-9tkm8p0g6r, 9tkm8p0g6r, p-ethylbenzoate, 4-ethyl benzoate, 4-ethylbenzoicacid, p-ethylbenzoic acid;
CCC1=CC=C(C=C1)C(=O)O
150.177
150.18

Chemical Identifiers

619-64-7
150.177
ZQVKTHRQIXSMGY-UHFFFAOYSA-N
12086
CCC1=CC=C(C=C1)C(=O)O
C9H10O2
MFCD00002570
benzoic acid, 4-ethyl, p-ethylbenzoic acid, 4-ethyl benzoic acid, 4-ethyl-benzoic acid, unii-9tkm8p0g6r, 9tkm8p0g6r, p-ethylbenzoate, 4-ethyl benzoate, 4-ethylbenzoicacid, p-ethylbenzoic acid;
4-ethylbenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents