missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Ethylbenzenethiol, TCI America™

Manufacturer:  TCI America E06725G

 View more versions of this product

Catalog No. E06725G



Specifications

Specifications

4-Ethylbenzenethiol
Yellow
4946-13-8
4-ethylthiophenol, 4-ethyl thiophenol, 4-ethylbenzene-1-thiol, benzenethiol, 4-ethyl, p-ethylphenylmercaptan, p-ethylthiophenol, pubchem6813, p-ethylphenyl mercaptan, 4-ethylthiophenol, acmc-209kg5
CCC1=CC=C(C=C1)S
138.228
138.23
≥97.0% (GC)
211°C
5g
C8H10S
WWQQPHUHTAZWDH-UHFFFAOYSA-N
4-ethylbenzenethiol
2758914
Clear Liquid at 20°C

Chemical Identifiers

4946-13-8
138.228
4-ethylthiophenol, 4-ethyl thiophenol, 4-ethylbenzene-1-thiol, benzenethiol, 4-ethyl, p-ethylphenylmercaptan, p-ethylthiophenol, pubchem6813, p-ethylphenyl mercaptan, 4-ethylthiophenol, acmc-209kg5
4-ethylbenzenethiol
C8H10S
WWQQPHUHTAZWDH-UHFFFAOYSA-N
2758914
CCC1=CC=C(C=C1)S
Safety and Handling

Safety and Handling

TSCA : No

Documents