missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Ethylbenzaldehyde, TCI America™

Manufacturer:  TCI America E0325100ML

 View more versions of this product

Catalog No. E0325100ML



Specifications

Specifications

4-Ethylbenzaldehyde
Colorless
4748-78-1
MFCD00006956
QNGNSVIICDLXHT-UHFFFAOYSA-N
4-ethylbenzaldehyde
20861
Clear Liquid at 20°C
221°C
100mL
C9H10O
p-ethylbenzaldehyde, benzaldehyde, 4-ethyl, 4-ethyl benzaldehyde, 4-ethyl-benzaldehyde, ebal, ethyl benzaldehyde, ethyl-benzaldehyde, unii-289ppw3sg4, benzaldehyde,4-ethyl, ethylbenzaldehyde, p
CCC1=CC=C(C=C1)C=O
134.178
134.18
≥98.0% (GC)

Chemical Identifiers

4748-78-1
134.178
QNGNSVIICDLXHT-UHFFFAOYSA-N
20861
CCC1=CC=C(C=C1)C=O
C9H10O
MFCD00006956
p-ethylbenzaldehyde, benzaldehyde, 4-ethyl, 4-ethyl benzaldehyde, 4-ethyl-benzaldehyde, ebal, ethyl benzaldehyde, ethyl-benzaldehyde, unii-289ppw3sg4, benzaldehyde,4-ethyl, ethylbenzaldehyde, p
4-ethylbenzaldehyde
Safety and Handling

Safety and Handling

EINECSNumber : (3)-3555

TSCA : Yes

Documents