Learn More
4-Ethylbenzaldehyde, 97%, Thermo Scientific Chemicals
$90.05 - $248.75
Chemical Identifiers
CAS | 4748-78-1 |
---|---|
Molecular Formula | C9H10O |
Molecular Weight (g/mol) | 134.178 |
MDL Number | MFCD00006956 |
InChI Key | QNGNSVIICDLXHT-UHFFFAOYSA-N |
Synonym | p-ethylbenzaldehyde, benzaldehyde, 4-ethyl, 4-ethyl benzaldehyde, 4-ethyl-benzaldehyde, ebal, ethyl benzaldehyde, ethyl-benzaldehyde, unii-289ppw3sg4, benzaldehyde,4-ethyl, ethylbenzaldehyde, p |
PubChem CID | 20861 |
IUPAC Name | 4-ethylbenzaldehyde |
SMILES | CCC1=CC=C(C=C1)C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2064514
|
Thermo Scientific Chemicals
B2064514 |
25 g |
Each for $90.05
|
|
|||||
AAB2064522
|
Thermo Scientific Chemicals
B2064522 |
100 g |
Each for $248.75
|
|
|||||
Description
It is used as a perfuming agent in cosmetics industry. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a perfuming agent in cosmetics industry. It is also used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.
Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Chemical Identifiers
4748-78-1 | |
134.178 | |
QNGNSVIICDLXHT-UHFFFAOYSA-N | |
20861 | |
CCC1=CC=C(C=C1)C=O |
C9H10O | |
MFCD00006956 | |
p-ethylbenzaldehyde, benzaldehyde, 4-ethyl, 4-ethyl benzaldehyde, 4-ethyl-benzaldehyde, ebal, ethyl benzaldehyde, ethyl-benzaldehyde, unii-289ppw3sg4, benzaldehyde,4-ethyl, ethylbenzaldehyde, p | |
4-ethylbenzaldehyde |
Specifications
4748-78-1 | |
220°C to 221°C | |
Characteristic | |
1.539 | |
25 g | |
Air Sensitive | |
Not miscible or difficult to mix in water. Soluble in alcohol. | |
CCC1=CC=C(C=C1)C=O | |
134.178 | |
134.18 | |
4-Ethylbenzaldehyde |
0.979 | |
92°C (197°F) | |
C9H10O | |
MFCD00006956 | |
636027 | |
p-ethylbenzaldehyde, benzaldehyde, 4-ethyl, 4-ethyl benzaldehyde, 4-ethyl-benzaldehyde, ebal, ethyl benzaldehyde, ethyl-benzaldehyde, unii-289ppw3sg4, benzaldehyde,4-ethyl, ethylbenzaldehyde, p | |
QNGNSVIICDLXHT-UHFFFAOYSA-N | |
4-ethylbenzaldehyde | |
20861 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
H227-H302
EINECSNumber : 225-268-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only