Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethylaniline, 98+%

Chemical Identifiers

Chemical Name or Material 4-Ethylaniline
CAS 589-16-2
Molecular Formula C8H11N
Formula Weight 121.18g/mol
MDL Number MFCD00007921
UN Number UN2810
Beilstein 774319
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2465222

 
alfa aesar™
B24652-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2465236

 
alfa aesar™
B24652-36
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point −5°C
Boiling Point 217° to 218°C
Quantity 100g
Flash Point 97°C (207°F)
Odor Characteristic
Chemical Name or Material 4-Ethylaniline
CAS 589-16-2
Assay ≥98%
Molecular Formula C8H11N
Formula Weight 121.18g/mol
MDL Number MFCD00007921
UN Number UN2810
Beilstein 774319
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H331-H373-H312
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P260-P261-P280-P301+P310-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P260-P261-P280-P301+P310-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H331-H373-H312
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.