Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Ethylacetophenone, 97%

$45.26 - $538.74

Chemical Identifiers

Chemical Name or Material 4'-Ethylacetophenone
CAS 937-30-4
Molecular Formula C10H12O
Formula Weight 148.21
MDL Number MFCD00009262
Beilstein 1906029
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2155714

 
alfa aesar™
B2155714
25g Each for $45.26

AAB2155722

 
alfa aesar™
B2155722
100g Each for $135.46

AAB2155736

 
alfa aesar™
B2155736
500g Each for $538.74
Description & Specifications

Specifications

Formula Weight 148.21
Chemical Name or Material 4'-Ethylacetophenone
TSCA TSCA
Assay 97%
Molecular Formula C10H12O
MDL Number MFCD00009262
CAS 937-30-4
Beilstein 1906029
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 90°C (194°F)
Boiling Point 239°C
Quantity 25g

Applications
It can be used to produce 1,1′-p-phenylene-bis-ethanone. This reaction will need reagent air, cobalt naphthenate. The reaction temperature is 125°C.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.