Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethyl-trans-beta-styrylboronic acid pinacol ester, 97%

$57.11 - $153.69

Chemical Identifiers

Chemical Name or Material 4-Ethyl-trans-beta-styrylboronic acid pinacol ester
CAS 870717-91-2
Molecular Formula C16H23BO2
Formula Weight 258.17
MDL Number MFCD08276837
Synonym 2-[trans-2-(4-Ethylphenyl)vinyl]-4,4,5,5-tetramethyl[1,3,2]dioxaborolane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3074403

  SDS
alfa aesar™
H3074403
1g Each for $153.69

AAH30744MD

  SDS
alfa aesar™
H30744MD
250mg Each for $57.11
Description & Specifications

Specifications

Formula Weight 258.17
Chemical Name or Material 4-Ethyl-trans-beta-styrylboronic acid pinacol ester
MDL Number MFCD08276837
CAS 870717-91-2
Assay 97%
Quantity 1g
Melting Point 46° to 48°C
Synonym 2-[trans-2-(4-Ethylphenyl)vinyl]-4,4,5,5-tetramethyl[1,3,2]dioxaborolane
Molecular Formula C16H23BO2
Flash Point >110°C (230°F)