Please login to your online account to display your discounted pricing

4-ethyl hydrogen 1-methyl-1H-pyrazole-4,5-dicarboxylate, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC76801CB

 View more versions of this product

Catalog No. CC76801CB

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 198.18
Quantity 250mg
Assay 97%
Packaging Amber Glass Bottle
Molecular Formula C8H10N2O4
Chemical Name or Material 4-ethyl hydrogen 1-methyl-1H-pyrazole-4, 5-dicarboxylate
CAS 81303-65-3

Specifications

Chemical Name or Material 4-ethyl hydrogen 1-methyl-1H-pyrazole-4, 5-dicarboxylate
Assay 97%
Molecular Formula C8H10N2O4
Formula Weight 198.18