Please login to your online account to display your discounted pricing

4-ethyl hydrogen 1-methyl-1H-pyrazole-4,5-dicarboxylate, Maybridge

$71.36 - $323.00

Chemical Identifiers

Chemical Name or Material 4-ethyl hydrogen 1-methyl-1H-pyrazole-4, 5-dicarboxylate
Assay 97%
Molecular Formula C8H10N2O4
Formula Weight 198.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC76801CB

  SDS
fisher scientific company
CC76801CB
Amber Glass Bottle 250mg Each for $71.36

CC76801DA

  SDS
fisher scientific company
CC76801DA
Amber Glass Bottle 1g Each for $323.00
Description & Specifications

Specifications

Formula Weight 198.18
Quantity 250mg
Assay 97%
Packaging Amber Glass Bottle
Molecular Formula C8H10N2O4
Chemical Name or Material 4-ethyl hydrogen 1-methyl-1H-pyrazole-4, 5-dicarboxylate
CAS 81303-65-3