missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Ethyl-4-methylmorpholinium Bromide 97.0+%, TCI America™
Supplier: TCI America E11665G
Specifications
4-Ethyl-4-methylmorpholinium Bromide | |
177°C | |
MFCD03095385 | |
ACQZVWGQFXXTIX-UHFFFAOYSA-M | |
4-ethyl-4-methylmorpholin-4-ium;bromide | |
103453 | |
≥97.0% (T) |
65756-41-4 | |
C7H16BrNO | |
5 g | |
CC[N+]1(CCOCC1)C.[Br-] | |
210.115 | |
210.12 | |
Crystalline Powder |
Chemical Identifiers
65756-41-4 | |
210.115 | |
ACQZVWGQFXXTIX-UHFFFAOYSA-M | |
4-ethyl-4-methylmorpholin-4-ium;bromide |
C7H16BrNO | |
MFCD03095385 | |
103453 | |
CC[N+]1(CCOCC1)C.[Br-] |
Safety and Handling
EINECSNumber : (5)-5971
TSCA : Yes