Please login to your online account to display your discounted pricing

4-Ethyl-4-formylhexanenitrile, 95%, ACROS Organics™

$123.09 - $123.09

Chemical Identifiers

Chemical Name or Material 4-Ethyl-4-formylhexanenitrile
Name Note 95%
CAS 2938-69-4
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C9H15NO
Linear Formula (C2H5)2C(CHO)CH2CH2CN
Formula Weight 153.22
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC195570050

  SDS
thermo fisher scientific
195570050
Glass bottle 5g Each for $123.09

AC195570250

  SDS
thermo fisher scientific
195570250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Liquid
Formula Weight 153.22
Quantity 5g
Color Clear colorless to light yellow
Density 0.948g/mL
EINECS Number 220-923-4
CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4505 to 1.4525 (20°C, 589nm)
Chemical Name or Material 4-Ethyl-4-formylhexanenitrile
Name Note 95%
Assay Percent Range 95%
Molecular Formula C9H15NO
Linear Formula (C2H5)2C(CHO)CH2CH2CN
MDL Number MFCD00047654
CAS 2938-69-4
Health Hazard 1 STENCH
Safety and Handling

STENCH