Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethyl-2-octanol, erythro + threo, 98%

$398.68 - $398.68

Chemical Identifiers

Chemical Name or Material 4-Ethyl-2-octanol
CAS 19780-78-0
Name Note erythro + threo
Molecular Formula C10H22O
Formula Weight 158.28
MDL Number MFCD00046660
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1995614

 
alfa aesar™
A1995614
25g Each for $398.68
Description & Specifications

Specifications

Formula Weight 158.28
Chemical Name or Material 4-Ethyl-2-octanol
Name Note erythro + threo
Assay 98%
Molecular Formula C10H22O
MDL Number MFCD00046660
CAS 19780-78-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.