Learn More
4-Ethyl-2-methoxyphenol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 351960250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Ethyl-2-methoxyphenol | |
2785-89-9 | |
100.0 | |
Colorless to Yellow | |
234°C to 236°C | |
Authentic | |
Glass bottle | |
1.5250 to 1.53 | |
25 g | |
4-ethylguaiacol, p-ethylguaiacol, 2-methoxy-4-ethylphenol, phenol, 4-ethyl-2-methoxy, homocresol, guaiacyl ethane, 4-ethyl-2-methoxy-phenol, 4-hydroxy-3-methoxy ethylbenzene, unii-c9nfd83bj5, guaiacol, 4-ethyl | |
CHWNEIVBYREQRF-UHFFFAOYSA-N | |
4-ethyl-2-methoxyphenol | |
62465 | |
98% |
98% | |
97.5 | |
15°C | |
1.0580g/mL | |
107°C | |
97.5% min. (GC) | |
C9H12O2 | |
MFCD00038714 | |
1.058 | |
Solubility in water: insoluble | |
CCC1=CC=C(O)C(OC)=C1 | |
152.19 | |
152.19 | |
Liquid |
Chemical Identifiers
2785-89-9 | |
152.19 | |
CHWNEIVBYREQRF-UHFFFAOYSA-N | |
62465 | |
CCC1=CC=C(O)C(OC)=C1 |
C9H12O2 | |
MFCD00038714 | |
4-ethylguaiacol, p-ethylguaiacol, 2-methoxy-4-ethylphenol, phenol, 4-ethyl-2-methoxy, homocresol, guaiacyl ethane, 4-ethyl-2-methoxy-phenol, 4-hydroxy-3-methoxy ethylbenzene, unii-c9nfd83bj5, guaiacol, 4-ethyl | |
4-ethyl-2-methoxyphenol |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
GHS Signal Word: Warning
EINECSNumber : 220-500-4
TSCA : TSCA
RUO – Research Use Only