Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethoxythiobenzamide, 97%

$108.18 - $272.73

Chemical Identifiers

Chemical Name or Material 4-Ethoxythiobenzamide
CAS 57774-79-5
Molecular Formula C9H11NOS
Formula Weight 181.25
MDL Number MFCD04629622
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2669103

  SDS
alfa aesar™
H2669103
1g Each for $272.73

AAH26691MD

  SDS
alfa aesar™
H26691MD
250mg Each for $108.18
Description & Specifications

Specifications

Formula Weight 181.25
Chemical Name or Material 4-Ethoxythiobenzamide
Assay 97%
Molecular Formula C9H11NOS
MDL Number MFCD04629622
CAS 57774-79-5
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 160° to 164°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.