Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethoxystyrene, 97%, stab. with 0.1% 4-tert-butylcatechol


Quantity 1g

Manufacturer: alfa aesar™  L0720903

 View more versions of this product

Catalog No. AAL0720903

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 148.21
Chemical Name or Material 4-Ethoxystyrene
CAS 5459-40-5
Assay 97%
MDL Number MFCD00009097
Health Hazard 1 Warning
Molecular Formula C10H12O
Recommended Storage 2° to 8°C
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Beilstein 1935209
Synonym 4-Vinylphenetole
Quantity 1g
Flash Point 87°C (188°F)
Boiling Point 99° to 100°C (11mmHg)
Name Note stabilized with 0.1% 4-tert-butylcatechol
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Specifications

Chemical Name or Material 4-Ethoxystyrene
CAS 5459-40-5
Name Note stabilized with 0.1% 4-tert-butylcatechol
Molecular Formula C10H12O
Formula Weight 148.21
MDL Number MFCD00009097
Beilstein 1935209
Synonym 4-Vinylphenetole
Accessories

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.

2° to 8°C

SDS & Safety Documents