Please login to your online account to display your discounted pricing

4-Ethoxyphenylboronic acid, 98%, ACROS Organics™

$46.92 - $46.92

Chemical Identifiers

Chemical Name or Material 4-Ethoxyphenylboronic acid
CAS 22237-13-4
CAS Min % 98.0
CAS Max % 100.0
Assay 98%
Assay Percent Range 98%
Molecular Formula C8H11BO3
Formula Weight 165.98
MDL Number MFCD00674028
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC359310050

  SDS
thermo fisher scientific
35931-0050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC359310010

  SDS
thermo fisher scientific
35931-0010
Glass bottle 1g Each for $46.92
Description & Specifications

Specifications

Formula Weight 165.98
Physical Form Crystalline Powder
Quantity 5g
Color White
Assay 98%
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C8H11BO3
Infrared Spectrum Authentic
MDL Number MFCD00674028
CAS 22237-13-4
Chemical Name or Material 4-Ethoxyphenylboronic acid
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.