Please login to your online account to display your discounted pricing

4-Ethoxyphenylboronic acid, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  35931-0010

 View more versions of this product

Catalog No. AC359310010

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 165.98
Quantity 1g
Color White
Assay 98%
CAS Max % 100.0
CAS Min % 98.0
CAS 22237-13-4
Assay Percent Range 98%
MDL Number MFCD00674028
Molecular Formula C8H11BO3
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 4-Ethoxyphenylboronic acid
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Ethoxyphenylboronic acid
CAS 22237-13-4
CAS Min % 98.0
CAS Max % 100.0
Assay 98%
Assay Percent Range 98%
Molecular Formula C8H11BO3
Formula Weight 165.98
MDL Number MFCD00674028
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

SDS & Safety Documents