missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Ethoxyphenylacetic acid, 98%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 156040250

 View more versions of this product

Catalog No. AC156040250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Ethoxyphenylacetic acid
Glass bottle
86°C to 90°C
98%
97.5
98%
C2H5OC6H4CH2CO2H
10, 190
ZVVWZNFSMIFGEP-UHFFFAOYSA-N
2-(4-ethoxyphenyl)acetic acid
78631
Crystalline Powder
Authentic
White to Beige
25g
4919-33-9
100.0
C10H12O3
MFCD00004346
2-4-ethoxyphenyl acetic acid
CCOC1=CC=C(C=C1)CC(=O)O
180.2
180.2
98%

Chemical Identifiers

4919-33-9
180.2
ZVVWZNFSMIFGEP-UHFFFAOYSA-N
78631
CCOC1=CC=C(C=C1)CC(=O)O
C10H12O3
MFCD00004346
2-4-ethoxyphenyl acetic acid
2-(4-ethoxyphenyl)acetic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 225-545-3

TSCA : TSCA

SDS Documents