missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Ethoxyphenylacetic Acid 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 156040100

 View more versions of this product

Catalog No. AC156040100

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
Crystalline Powder
White to beige
10, 190
Authentic
Glass bottle
4919-33-9
C2H5OC6H4CH2CO2H
98%
180.2
10g
86° to 90°C
100.0
97.5
MFCD00004346
4-Ethoxyphenylacetic acid
C10H12O3
98%

Chemical Identifiers

4919-33-9
100.0
C10H12O3
180.2
4-Ethoxyphenylacetic acid, 98%
97.5
98%
C2H5OC6H4CH2CO2H
MFCD00004346
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 225-545-3

TSCA : TSCA