Please login to your online account to display your discounted pricing

4-Ethoxyphenylacetic Acid 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Ethoxyphenylacetic acid
Name Note 98%
CAS 4919-33-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H12O3
Linear Formula C2H5OC6H4CH2CO2H
Formula Weight 180.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC156040100

  SDS
thermo fisher scientific
156040100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC156040250

  SDS
thermo fisher scientific
156040250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 180.2
Quantity 10g
Color White to beige
EINECS Number 225-545-3
TSCA TSCA
Melting Point 86° to 90°C
Beilstein 10, 190
CAS Max % 100.0
Packaging Glass bottle
MDL Number MFCD00004346
CAS 4919-33-9
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material 4-Ethoxyphenylacetic acid
Name Note 98%
Assay Percent Range 98%
Molecular Formula C10H12O3
Linear Formula C2H5OC6H4CH2CO2H
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.