missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethoxyphenyl 4-n-butylcyclohexanecarboxylate, 97%

$94.93 - $378.66

Chemical Identifiers

CAS 67589-47-3
Molecular Formula C19H28O3
Molecular Weight (g/mol) 304.43
MDL Number MFCD00409324
InChI Key JAYBMJPZPJBTQZ-UHFFFAOYSA-N
Synonym 4-ethoxyphenyl 4-butylcyclohexanecarboxylate
PubChem CID 105459
IUPAC Name (4-ethoxyphenyl) 4-butylcyclohexane-1-carboxylate
SMILES CCCCC1CCC(CC1)C(=O)OC2=CC=C(C=C2)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5553703 View Documents Alfa Aesar™
H5553703
1g Each for $94.93
 
AAH5553706 View Documents Alfa Aesar™
H5553706
5g Each for $378.66
 
Specifications

Chemical Identifiers

67589-47-3
304.43
JAYBMJPZPJBTQZ-UHFFFAOYSA-N
105459
CCCCC1CCC(CC1)C(=O)OC2=CC=C(C=C2)OCC
C19H28O3
MFCD00409324
4-ethoxyphenyl 4-butylcyclohexanecarboxylate
(4-ethoxyphenyl) 4-butylcyclohexane-1-carboxylate

Specifications

35°C to 38°C
67589-47-3
MFCD00409324
JAYBMJPZPJBTQZ-UHFFFAOYSA-N
(4-ethoxyphenyl) 4-butylcyclohexane-1-carboxylate
105459
97%
1g
C19H28O3
4-ethoxyphenyl 4-butylcyclohexanecarboxylate
CCCCC1CCC(CC1)C(=O)OC2=CC=C(C=C2)OCC
304.43
304.43
4-Ethoxyphenyl 4-n-butylcyclohexanecarboxylate
Safety and Handling

Safety and Handling

EINECSNumber : 266-745-0

TSCA : Yes

Recommended Storage : Ambient temperatures