missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethoxyphenyl 4-n-butylcyclohexanecarboxylate, 97%

Manufacturer:  Alfa Aesar™ H5553706

 View more versions of this product

Catalog No. AAH5553706



Specifications

Specifications

4-Ethoxyphenyl 4-n-butylcyclohexanecarboxylate
5g
C19H28O3
4-ethoxyphenyl 4-butylcyclohexanecarboxylate
CCCCC1CCC(CC1)C(=O)OC2=CC=C(C=C2)OCC
304.43
304.43
35°C to 38°C
67589-47-3
MFCD00409324
JAYBMJPZPJBTQZ-UHFFFAOYSA-N
(4-ethoxyphenyl) 4-butylcyclohexane-1-carboxylate
105459
97%

Chemical Identifiers

67589-47-3
304.43
JAYBMJPZPJBTQZ-UHFFFAOYSA-N
105459
CCCCC1CCC(CC1)C(=O)OC2=CC=C(C=C2)OCC
C19H28O3
MFCD00409324
4-ethoxyphenyl 4-butylcyclohexanecarboxylate
(4-ethoxyphenyl) 4-butylcyclohexane-1-carboxylate
Safety and Handling

Safety and Handling

EINECSNumber : 266-745-0

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents