Please login to your online account to display your discounted pricing

4-Ethoxyphenol, 99%, ACROS Organics™

$71.36 - $71.36

Chemical Identifiers

Chemical Name or Material 4-Ethoxyphenol
CAS 622-62-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H10O2
Formula Weight 138.17
MDL Number MFCD00002334
Synonym Hydroquinone monoethyl ether
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC409671000

  SDS
thermo fisher scientific
AC409671000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC409670250

  SDS
thermo fisher scientific
409670250
Glass bottle 25g Each for $71.36
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 138.17
Physical Form Crystalline Powder or Chunks
Quantity 100g
Color Beige
Melting Point 64° to 67°C
CAS Max % 100.0
CAS 622-62-8
Chemical Name or Material 4-Ethoxyphenol
Molecular Formula C8H10O2
Assay Percent Range 99%
CAS Min % 98.5
MDL Number MFCD00002334
Packaging Glass bottle
Synonym Hydroquinone monoethyl ether
Infrared Spectrum Authentic
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Danger

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.