missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethoxymethylene-2-phenyloxazolin-5-one, 98+%

Manufacturer:  Alfa Aesar™ A1015506

 View more versions of this product

Catalog No. AAA1015506



Description

Description

Applications
4-Ethoxymethylene-2-phenyloxazolin-5-one acts as sensitizing agent. It is also used as pharmaceutical intermediates.

Solubility
Soluble in methanol (50 mg/ml), and water (partly).

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
Specifications

Specifications

4-Ethoxymethylene-2-phenyloxazolin-5-one
5g
C12H11NO3
163055
SJHPCNCNNSSLPL-NTMALXAHSA-N
(4Z)-4-(ethoxymethylidene)-2-phenyl-1,3-oxazol-5-one
1712094
≥98%
93°C to 97°C
15646-46-5
MFCD00003204
oxazolone, oxazolone sensitizing agent, 5 4h-oxazolone, 4-ethoxymethylene-2-phenyl, unii-78il0j7n72, 4-ethoxymethylene-2-phenyl-2-oxazolin-5-one, 4-ethoxymethylene-2-phenyloxazolin-5-one, 4-ethoxymethylene-2-phenyl-2-oxazoline-5-one, 4z-4-ethoxymethylidene-2-phenyl-1,3-oxazol-5-one, 2-oxazolin-5-one, 4-ethoxymethylene-2-phenyl, 4-ethoxymethylene-2-phenyloxazolone
CCOC=C1C(=O)OC(=N1)C2=CC=CC=C2
217.224
217.22

Chemical Identifiers

15646-46-5
217.224
SJHPCNCNNSSLPL-NTMALXAHSA-N
1712094
CCOC=C1C(=O)OC(=N1)C2=CC=CC=C2
C12H11NO3
MFCD00003204
oxazolone, oxazolone sensitizing agent, 5 4h-oxazolone, 4-ethoxymethylene-2-phenyl, unii-78il0j7n72, 4-ethoxymethylene-2-phenyl-2-oxazolin-5-one, 4-ethoxymethylene-2-phenyloxazolin-5-one, 4-ethoxymethylene-2-phenyl-2-oxazoline-5-one, 4z-4-ethoxymethylidene-2-phenyl-1,3-oxazol-5-one, 2-oxazolin-5-one, 4-ethoxymethylene-2-phenyl, 4-ethoxymethylene-2-phenyloxazolone
(4Z)-4-(ethoxymethylidene)-2-phenyl-1,3-oxazol-5-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : RQ5786250

Recommended Storage : 2°C to 8°C

SDS Documents