Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Ethoxymethyl)phenylacetic acid

$346.34 - $346.34

Chemical Identifiers

Chemical Name or Material 4-(Ethoxymethyl)phenylacetic acid
Molecular Formula C11H14O3
Formula Weight 194.23
MDL Number MFCD08061030
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5085503

 
alfa aesar™
H5085503
1g Each for $346.34

AAH50855MD

 
alfa aesar™
H50855MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 194.23
Chemical Name or Material 4-(Ethoxymethyl)phenylacetic acid
Molecular Formula C11H14O3
MDL Number MFCD08061030
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.