missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Ethoxycoumarin, TCI America™

Manufacturer:  TCI America E054525G

 View more versions of this product

Catalog No. E054525G



Specifications

Specifications

4-Ethoxycoumarin
137°C
35817-27-7
MFCD00052755
CCOC1=CC(=O)OC2=CC=CC=C21
190.198
190.20
≥98.0% (GC)
Yellow
25g
C11H10O3
FLZDKBHAUILYAZ-UHFFFAOYSA-N
4-ethoxychromen-2-one
688703
Crystal-Powder at 20°C

Chemical Identifiers

35817-27-7
190.198
FLZDKBHAUILYAZ-UHFFFAOYSA-N
4-ethoxychromen-2-one
C11H10O3
MFCD00052755
688703
CCOC1=CC(=O)OC2=CC=CC=C21
Safety and Handling

Safety and Handling

TSCA : No

Documents