Please login to your online account to display your discounted pricing

4-Ethoxycarbonyloxy-3,5-dimethoxybenzoic acid, 96%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  183500050

 View more versions of this product

Catalog No. AC183500050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Powder
Formula Weight 270.23
Quantity 5g
Color Beige
Melting Point 180° to 185°C
EINECS Number 242-567-9
Beilstein 10, I, 241
CAS Min % 95.0
CAS Max % 100.0
Chemical Name or Material 4-Ethoxycarbonyloxy-3, 5-dimethoxybenzoic acid
Packaging Glass bottle
Name Note 96%
Assay Percent Range 96%
Molecular Formula C12H14O7
Linear Formula C2H5OCO2C6H2(OCH3)2CO2H
MDL Number MFCD00002503
CAS 18780-67-1
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 4-Ethoxycarbonyloxy-3, 5-dimethoxybenzoic acid
Name Note 96%
CAS 18780-67-1
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C12H14O7
Linear Formula C2H5OCO2C6H2(OCH3)2CO2H
Formula Weight 270.23
MDL Number MFCD00002503