Please login to your online account to display your discounted pricing

4-Ethoxycarbonyloxy-3,5-dimethoxybenzoic acid, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Ethoxycarbonyloxy-3, 5-dimethoxybenzoic acid
Name Note 96%
CAS 18780-67-1
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C12H14O7
Linear Formula C2H5OCO2C6H2(OCH3)2CO2H
Formula Weight 270.23
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC183500050

  SDS
thermo fisher scientific
183500050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95%
Physical Form Powder
Formula Weight 270.23
Quantity 5g
Color Beige
Melting Point 180° to 185°C
EINECS Number 242-567-9
Beilstein 10, I, 241
CAS Min % 95.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 96%
Assay Percent Range 96%
CAS 18780-67-1
Linear Formula C2H5OCO2C6H2(OCH3)2CO2H
Molecular Formula C12H14O7
Chemical Name or Material 4-Ethoxycarbonyloxy-3, 5-dimethoxybenzoic acid
MDL Number MFCD00002503