Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Ethoxycarbonylamino)piperidine, 97%

Chemical Identifiers

Chemical Name or Material 4-(Ethoxycarbonylamino)piperidine
CAS 64951-36-6
Formula Weight 172.22
MDL Number MFCD03844679
Synonym Ethyl 4-piperidinylcarbamate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6309503

 
alfa aesar™
H6309503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6309506

 
alfa aesar™
H6309506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63095MD

 
alfa aesar™
H63095MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 172.22
Chemical Name or Material 4-(Ethoxycarbonylamino)piperidine
Physical Form Solid
Assay 97%
MDL Number MFCD03844679
CAS 64951-36-6
Synonym Ethyl 4-piperidinylcarbamate
Quantity 1g