Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Ethoxycarbonyl)phenyl isocyanate, 97%


Quantity 5g

Manufacturer: alfa aesar™  L1068106

 View more versions of this product

Catalog No. AAL1068106

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 191.19
CAS 30806-83-8
Assay 97%
Molecular Formula C10H9NO3
Chemical Name or Material 4-(Ethoxycarbonyl)phenyl isocyanate
MDL Number MFCD00002027
UN Number UN2811
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Flash Point >110°C (230°F)
Melting Point 28° to 32°C
Beilstein 882021
Synonym 4-Carboethoxyphenyl isocyanate; Ethyl 4-isocyanatobenzoate
Health Hazard 1 Danger
Recommended Storage 2° to 8°C
Boiling Point 118° to 119°C (0.8mmHg)
Quantity 5g

Specifications

Chemical Name or Material 4-(Ethoxycarbonyl)phenyl isocyanate
CAS 30806-83-8
Molecular Formula C10H9NO3
Formula Weight 191.19
MDL Number MFCD00002027
UN Number UN2811
Beilstein 882021
Synonym 4-Carboethoxyphenyl isocyanate; Ethyl 4-isocyanatobenzoate

Applications
Ethyl 4-isocyanatobenzoate was used in the preparation of cellulose carbamate and ester derivatives. It was also used in the preparation of ethyl 4-(3-(4-oxo-6-tridecyl-1,4-dihydropyrimidin-2-yl)ureido)benzoate2 and ethyl 4-(2-oxocyclohexanecarboxamido)benzoate.

Solubility
Hydrolyses in water.

Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from moisture, alocohol, amines, water.

Accessories

Danger

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C