missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-[(Ethoxycarbonyl)methyl]phenylboronic acid pinacol ester, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC445000010

 View more versions of this product

Catalog No. AC445000010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-[(Ethoxycarbonyl)methyl]phenylboronic acid pinacol ester
41°C to 43°C
859169-20-3
C16H23BO4
RPRBNRDIESZHFL-UHFFFAOYSA-N
ethyl 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate
46737998
97%
Glass Bottle
1g
96% min. (GC)
4-2-ethoxy-2-oxoethyl phenyl boronic acid pinacol ester, 4-ethoxycarbonylmethyl phenylboronic acid pinacol ester, 4-ethoxycarbonylmethyl phenylboronic acid, pinacol ester, 4-ethoxycarbonylmethylphenyl boronic acid pinacol ester, 4-ethoxycarbonylmethylphenyl boronic acid, pinacol ester, 4-ethoxycarbonylmethylphenylboronic acid pinacol ester, 4-ethoxycarbonylmethylphenylboronic acid, pinacol ester, ethyl 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, ethyl 2-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CC(=O)OCC
290.16
290.16

Chemical Identifiers

859169-20-3
290.16
4-2-ethoxy-2-oxoethyl phenyl boronic acid pinacol ester, 4-ethoxycarbonylmethyl phenylboronic acid pinacol ester, 4-ethoxycarbonylmethyl phenylboronic acid, pinacol ester, 4-ethoxycarbonylmethylphenyl boronic acid pinacol ester, 4-ethoxycarbonylmethylphenyl boronic acid, pinacol ester, 4-ethoxycarbonylmethylphenylboronic acid pinacol ester, 4-ethoxycarbonylmethylphenylboronic acid, pinacol ester, ethyl 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, ethyl 2-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate
ethyl 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate
C16H23BO4
RPRBNRDIESZHFL-UHFFFAOYSA-N
46737998
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CC(=O)OCC
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS Documents