Please login to your online account to display your discounted pricing

Alfa Aesar™ [4-(Ethoxycarbonyl)butyl]triphenylphosphonium bromide, 95%

$61.03 - $230.54

Chemical Identifiers

Chemical Name or Material [4-(Ethoxycarbonyl)butyl]triphenylphosphonium bromide
CAS 54110-96-2
Molecular Formula C25H28BrO2P
Linear Formula CH3CH2O2C(CH2)4P(C6H5)3Br
Formula Weight 471.38
MDL Number MFCD00051866
Beilstein 5212054
Synonym [4-(Carboethoxy)butyl]triphenylphosphonium bromide; Ethyl 4-(triphenylphosphonio)butyrate bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0591114

  SDS
alfa aesar™
L0591114
25g Each for $230.54

AAL0591106

  SDS
alfa aesar™
L0591106
5g Each for $61.03
Description & Specifications

Specifications

Chemical Name or Material [4-(Ethoxycarbonyl)butyl]triphenylphosphonium bromide
Quantity 25g
CAS 54110-96-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Assay 95%
Molecular Formula C25H28BrO2P
Linear Formula CH3CH2O2C(CH2)4P(C6H5)3Br
Formula Weight 471.38
MDL Number MFCD00051866
Beilstein 5212054
Synonym [4-(Carboethoxy)butyl]triphenylphosphonium bromide; Ethyl 4-(triphenylphosphonio)butyrate bromide

Applications
[4-(Ethoxycarbonyl)butyl]triphenylphosphonium bromide is used as catalytic agent, petrochemical additive.

Notes
Store under dry inert gas. Protect from humidity and water. Incompatible with moisture and oxidizing agents. Hygroscopic.

Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.