missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, 97%

$49.68 - $421.08

Chemical Identifiers

CAS 195062-62-5
Molecular Formula C15H21BO4
Molecular Weight (g/mol) 276.139
MDL Number MFCD03453659
InChI Key NCVIYKCFTYSAGN-UHFFFAOYSA-N
Synonym 2-4-ethoxycarbonyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 2734618
IUPAC Name ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)OCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH55836MD View Documents Alfa Aesar™
H55836MD
250mg Each for $49.68
 
AAH5583603 View Documents Alfa Aesar™
H5583603
1g Each for $130.34
 
AAH5583606 View Documents Alfa Aesar™
H5583606
5g Each for $421.08
 
Specifications

Chemical Identifiers

195062-62-5
276.139
NCVIYKCFTYSAGN-UHFFFAOYSA-N
2734618
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)OCC
C15H21BO4
MFCD03453659
2-4-ethoxycarbonyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

Specifications

1.052g/mL
330°C
250mg
C15H21BO4
2-4-ethoxycarbonyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)OCC
276.139
276.14
97%
1.5000
Colorless
195062-62-5
MFCD03453659
NCVIYKCFTYSAGN-UHFFFAOYSA-N
ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
2734618
Liquid
4-(Ethoxycarbonyl)benzeneboronic acid pinacol ester