missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, 97%

Manufacturer:  Alfa Aesar™ H55836MD

 View more versions of this product

Catalog No. AAH55836MD



Specifications

Specifications

4-(Ethoxycarbonyl)benzeneboronic acid pinacol ester
1.5000
Colorless
195062-62-5
MFCD03453659
NCVIYKCFTYSAGN-UHFFFAOYSA-N
ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
2734618
Liquid
1.052g/mL
330°C
250mg
C15H21BO4
2-4-ethoxycarbonyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)OCC
276.139
276.14
97%

Chemical Identifiers

195062-62-5
276.139
NCVIYKCFTYSAGN-UHFFFAOYSA-N
2734618
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)OCC
C15H21BO4
MFCD03453659
2-4-ethoxycarbonyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Documents