missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester, 97%

$432.76 - $1,345.36

Chemical Identifiers

CAS 851334-92-4
Molecular Formula C15H20BFO4
Molecular Weight (g/mol) 294.129
MDL Number MFCD18383988
InChI Key ZXSGWPHDXQZIAF-UHFFFAOYSA-N
Synonym 4-ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester
PubChem CID 23134230
IUPAC Name ethyl 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C(=O)OCC)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH55087MD View Documents Alfa Aesar™
H55087MD
250mg Each for $432.76
 
AAH5508703 View Documents Alfa Aesar™
H5508703
1g Each for $1,345.36
 
Specifications

Chemical Identifiers

851334-92-4
294.129
ZXSGWPHDXQZIAF-UHFFFAOYSA-N
23134230
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C(=O)OCC)F
C15H20BFO4
MFCD18383988
4-ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester
ethyl 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

Specifications

250mg
C15H20BFO4
14562502
ZXSGWPHDXQZIAF-UHFFFAOYSA-N
ethyl 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
23134230
97%
851334-92-4
MFCD18383988
4-ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C(=O)OCC)F
294.129
294.13
4-Ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures