missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester, 97%

Manufacturer:  Alfa Aesar™ H55087MD

 View more versions of this product

Catalog No. AAH55087MD



Description

Description

Specifications

Specifications

4-Ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester
851334-92-4
MFCD18383988
4-ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester, 4-ethoxycarbonyl-2-fluorophenylboronic acid, pinacol ester, ethyl 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzoate, ethyl 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, ethyl 3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, ethyl3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C(=O)OCC)F
294.129
294.13
250mg
C15H20BFO4
14562502
ZXSGWPHDXQZIAF-UHFFFAOYSA-N
ethyl 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
23134230
97%

Chemical Identifiers

851334-92-4
294.129
ZXSGWPHDXQZIAF-UHFFFAOYSA-N
23134230
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C(=O)OCC)F
C15H20BFO4
MFCD18383988
4-ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester, 4-ethoxycarbonyl-2-fluorophenylboronic acid, pinacol ester, ethyl 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzoate, ethyl 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, ethyl 3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, ethyl3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate
ethyl 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents