Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Ethoxybiphenyl-4-boronic acid, 97%

Chemical Identifiers

Chemical Name or Material 4'-Ethoxybiphenyl-4-boronic acid
CAS 182344-29-2
Formula Weight 242.08
MDL Number MFCD06798054
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH55859MD

 
alfa aesar™
H55859MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5585903

 
alfa aesar™
H5585903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 242.08
Chemical Name or Material 4'-Ethoxybiphenyl-4-boronic acid
Physical Form Solid
Assay 97%
MDL Number MFCD06798054
CAS 182344-29-2
Quantity 250mg