Alfa Aesar™ 4'-Ethoxybiphenyl-4-boronic acid, 97%
Manufacturer:  Alfa Aesar™ H55859MD
 View more versions of this product
Catalog No. AAH55859MD
Specifications
Specifications
4'-Ethoxybiphenyl-4-boronic acid
182344-29-2
MFCD06798054
ZFFXATWYKJBSCC-UHFFFAOYSA-N
[4-(4-ethoxyphenyl)phenyl]boronic acid
16217237
Solid
250mg
C14H15BO3
4'-ethoxy 1,1'-biphenyl-4-yl boronic acid
B(C1=CC=C(C=C1)C2=CC=C(C=C2)OCC)(O)O
242.081
242.08
97%
Chemical Identifiers
182344-29-2
242.081
ZFFXATWYKJBSCC-UHFFFAOYSA-N
16217237
B(C1=CC=C(C=C1)C2=CC=C(C=C2)OCC)(O)O
C14H15BO3
MFCD06798054
4'-ethoxy 1,1'-biphenyl-4-yl boronic acid
[4-(4-ethoxyphenyl)phenyl]boronic acid
Safety and Handling
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
Documents