missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Ethoxybenzyltriphenylphosphonium Bromide, TCI America™

Manufacturer:  TCI America E042125G

 View more versions of this product

Catalog No. E042125G



Specifications

Specifications

4-Ethoxybenzyltriphenylphosphonium Bromide
225°C
82105-88-2
MFCD00011834
MZBKKJRCQNVENM-UHFFFAOYSA-M
(4-ethoxyphenyl)methyl-triphenylphosphanium;bromide
2724470
Crystal-Powder at 20°C
White-Yellow
25g
C27H26BrOP
4-ethoxybenzyltriphenylphosphonium bromide, 4-ethoxybenzyl triphenylphosphonium bromide, 4-ethoxyphenyl methyl triphenylphosphanium bromide, acmc-209pn2, 4-ethoxybenzyltriphenylphosphoniumbromide, p-ethoxybenzyltriphenylphosphonium bromide, 4-ethoxybenzyl triphenylphosphoniumbromide, 4-ethoxybenzyl triphenyl phosphonium bromide, 4-ethoxyphenyl methyl-triphenylphosphanium bromide, 4-ethoxyphenyl methyl triphenyl phosphanium bromide
CCOC1=CC=C(C=C1)C[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[Br-]
477.382
477.38
≥98.0% (T)

Chemical Identifiers

82105-88-2
477.382
MZBKKJRCQNVENM-UHFFFAOYSA-M
2724470
CCOC1=CC=C(C=C1)C[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[Br-]
C27H26BrOP
MFCD00011834
4-ethoxybenzyltriphenylphosphonium bromide, 4-ethoxybenzyl triphenylphosphonium bromide, 4-ethoxyphenyl methyl triphenylphosphanium bromide, acmc-209pn2, 4-ethoxybenzyltriphenylphosphoniumbromide, p-ethoxybenzyltriphenylphosphonium bromide, 4-ethoxybenzyl triphenylphosphoniumbromide, 4-ethoxybenzyl triphenyl phosphonium bromide, 4-ethoxyphenyl methyl-triphenylphosphanium bromide, 4-ethoxyphenyl methyl triphenyl phosphanium bromide
(4-ethoxyphenyl)methyl-triphenylphosphanium;bromide
Safety and Handling

Safety and Handling

TSCA : No

Documents