missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4′-Ethoxybenzylidene-4-butylaniline, TCI America™

Manufacturer:  TCI America E026225G

 View more versions of this product

Catalog No. E026225G



Specifications

Specifications

4′-Ethoxybenzylidene-4-butylaniline
25g
C19H23NO
EBBA
CCCCC1=CC=C(C=C1)N=CC2=CC=C(C=C2)OCC
281.399
281.40
≥99.0% (T)
White-Yellow
29743-08-6
MFCD00009465
DBOAVDSSZWDGTH-UHFFFAOYSA-N
N-(4-butylphenyl)-1-(4-ethoxyphenyl)methanimine
94406
Crystal-Powder at 20°C

Chemical Identifiers

29743-08-6
281.399
DBOAVDSSZWDGTH-UHFFFAOYSA-N
94406
CCCCC1=CC=C(C=C1)N=CC2=CC=C(C=C2)OCC
C19H23NO
MFCD00009465
EBBA
N-(4-butylphenyl)-1-(4-ethoxyphenyl)methanimine
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1218

TSCA : Yes

Recommended Storage : Refrigerator

Documents