Please login to your online account to display your discounted pricing

4-Ethoxybenzoic acid, 99%, ACROS Organics™


Quantity 500g

Manufacturer: thermo fisher scientific  163255000

 View more versions of this product

Catalog No. AC163255000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Very Fine Crystalline Powder
Assay ≥98.5%
Formula Weight 166.18
Color White
Quantity 500g
EINECS Number 210-616-3
TSCA TSCA
Melting Point 197° to 199°C
Beilstein 10, 156
CAS Max % 100.0
Chemical Name or Material 4-Ethoxybenzoic acid
Name Note 99%
Assay Percent Range 99%
Molecular Formula C9H10O3
Linear Formula C2H5OC6H4CO2H
Infrared Spectrum Authentic
MDL Number MFCD00002545
CAS 619-86-3
CAS Min % 98.5

Specifications

Chemical Name or Material 4-Ethoxybenzoic acid
Name Note 99%
CAS 619-86-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H10O3
Linear Formula C2H5OC6H4CO2H
Formula Weight 166.18
MDL Number MFCD00002545