Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethoxybenzhydrazide, 98+%

$105.89 - $105.89

Chemical Identifiers

Chemical Name or Material 4-Ethoxybenzhydrazide
CAS 58586-81-5
Molecular Formula C9H12N2O2
Formula Weight 180.21
MDL Number MFCD00017074
Beilstein 2642900
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0543306

  SDS
alfa aesar™
L0543306
5g Each for $105.89
Description & Specifications

Specifications

Formula Weight 180.21
Chemical Name or Material 4-Ethoxybenzhydrazide
Molecular Formula C9H12N2O2
Assay ≥98%
MDL Number MFCD00017074
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 58586-81-5
Beilstein 2642900
Health Hazard 1 Warning
Melting Point 124° to 127°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.