missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Ethoxybenzaldehyde, 97+%

Manufacturer:  Alfa Aesar™ A1198722

 View more versions of this product

Catalog No. AAA1198722


 

Description

Description

Applications
4-Ethoxybenzaldehyde can be used as an anti-inflammatory and prostaglandin E2 suppressor. benzaldehyde-derived anti-inflammatory agents may be useful in reducing facial erythema.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents. Air sensitive.
Specifications

Specifications

4-Ethoxybenzaldehyde
≥97%
MFCD00003388
JRHHJNMASOIRDS-UHFFFAOYSA-N
4-ethoxybenzaldehyde
24834
253°C to 255°C
100g
10031-82-0
C9H10O2
386863
CCOC1=CC=C(C=C1)C=O
150.177
150.18
13°C to 14°C
<110°C (230°F)

Chemical Identifiers

10031-82-0
150.177
JRHHJNMASOIRDS-UHFFFAOYSA-N
C9H10O2
MFCD00003388
24834
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

RTECSNumber : CU6100000

TSCA : TSCA

SDS Documents