missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4'-Ethoxyacetanilide, 97%, Thermo Scientific Chemicals
$53.19 - $145.71
Chemical Identifiers
CAS | 62-44-2 |
---|---|
Molecular Formula | C10H13NO2 |
Molecular Weight (g/mol) | 179.219 |
MDL Number | MFCD00009094 |
InChI Key | CPJSUEIXXCENMM-UHFFFAOYSA-N |
Synonym | phenacetin, acetophenetidin, n-4-ethoxyphenyl acetamide, acetphenetidin, acetophenetidine, acetophenetin, phenacetine, p-acetophenetidide, contradouleur, achrocidin |
PubChem CID | 4754 |
ChEBI | CHEBI:8050 |
IUPAC Name | N-(4-ethoxyphenyl)acetamide |
SMILES | CCOC1=CC=C(C=C1)NC(=O)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1120022
|
Thermo Scientific Chemicals
A1120022 |
100 g |
N/A
|
|
|||||
AAA1120036
|
Thermo Scientific Chemicals
A1120036 |
500 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
62-44-2 | |
179.219 | |
CPJSUEIXXCENMM-UHFFFAOYSA-N | |
4754 | |
N-(4-ethoxyphenyl)acetamide |
C10H13NO2 | |
MFCD00009094 | |
phenacetin, acetophenetidin, n-4-ethoxyphenyl acetamide, acetphenetidin, acetophenetidine, acetophenetin, phenacetine, p-acetophenetidide, contradouleur, achrocidin | |
CHEBI:8050 | |
CCOC1=CC=C(C=C1)NC(=O)C |
Specifications
62-44-2 | |
C10H13NO2 | |
100 g | |
14,7204 | |
CPJSUEIXXCENMM-UHFFFAOYSA-N | |
N-(4-ethoxyphenyl)acetamide | |
4754 | |
179.22 | |
4'-Ethoxyacetanilide |
134°C to 136°C | |
MFCD00009094 | |
1869238 | |
phenacetin, acetophenetidin, n-4-ethoxyphenyl acetamide, acetphenetidin, acetophenetidine, acetophenetin, phenacetine, p-acetophenetidide, contradouleur, achrocidin | |
CCOC1=CC=C(C=C1)NC(=O)C | |
179.219 | |
CHEBI:8050 | |
97% |
Safety and Handling
GHS H Statement
H350-H302
May cause cancer.
Harmful if swallowed.
P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
H302-H350
EINECSNumber : 200-533-0
RTECSNumber : AM4375000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only